siRNA Details
Gene Symbol: BTF3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136497.2 | NP_001129969.1 | 5 | 1878 | ||
NM_001243767.1 | NP_001230696.1 | 5 | 1743 | |||
NM_152265.4 | NP_689478.1 | 6 | 1945 | |||
GenBank mRNA | AK027582.1 | - | 5 | 1760 | ||
AK027750.1 | - | 6 | 1827 | |||
BC021004.1 | - | 5 | 1722 | |||
BC022371.1 | - | 6 | 1806 | 4739327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
