siRNA Details
Gene Symbol: TSPAN32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139022.2 | NP_620591.3 | 10 | 1256 | ||
XM_005252719.4 | XP_005252776.1 | 9 | 1587 | |||
XM_011519815.2 | XP_011518117.2 | 8 | 1509 | |||
XM_011519818.2 | XP_011518120.2 | 9 | 1172 | |||
XM_011519819.1 | XP_011518121.1 | 9 | 1475 | |||
XM_011519822.1 | XP_011518124.1 | 9 | 1068 | |||
XM_017017064.1 | XP_016872553.1 | 8 | 2059 | |||
XM_017017065.1 | XP_016872554.1 | 9 | 1722 | |||
XM_017017066.1 | XP_016872555.1 | 7 | 1975 | |||
XM_017017067.1 | XP_016872556.1 | 9 | 1479 | |||
XM_017017068.1 | XP_016872557.1 | 8 | 1540 | |||
GenBank mRNA | AF125569.1 | - | 9 | 1174 | ||
AF176070.1 | - | 9 | 1140 | |||
AF176071.1 | - | 8 | 842 | |||
AK128812.1 | - | 9 | 3220 | |||
AY039001.1 | - | 10 | 1125 | |||
BC016693.1 | - | 10 | 1204 | 4071111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
