siRNA Details
Gene Symbol: NANOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199461.3 | NP_955631.1 | 1 | 1134 | ||
GenBank mRNA | AF275269.2 | - | 1 | 1090 | ||
AF458985.1 | - | 1 | 1134 | |||
AI276145.1 | - | 2 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
