siRNA Details
Gene Symbol: PDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002597.4 | NP_002588.3 | 4 | 357 | ||
NM_022576.3 | NP_072098.1 | 3 | 319 | |||
XM_011509603.2 | XP_011507905.1 | 3 | 1375 | |||
GenBank mRNA | AF076464.1 | - | 3 | 320 | ||
BC093839.1 | - | 2 | 150 | |||
BC112134.1 | - | 2 | 150 | |||
EL945964.1 | - | 4 | 361 | |||
M33478.1 | - | 4 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
