siRNA Details
Gene Symbol: PDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002597.4 | NP_002588.3 | 4 | 827 | ||
NM_022576.3 | NP_072098.1 | 3 | 789 | |||
XM_011509603.2 | XP_011507905.1 | 3 | 1845 | |||
GenBank mRNA | AF076464.1 | - | 3 | 790 | ||
BC093839.1 | - | 2 | 620 | |||
BC112134.1 | - | 2 | 620 | |||
BM715757.1 | - | 1 | 403 | |||
M33478.1 | - | 4 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
