siRNA Details
Gene Symbol: SPAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039592.1 | NP_001034681.1 | 2 | 917 | ||
NM_172312.1 | NP_758516.1 | 2 | 917 | |||
GenBank mRNA | AK302313.1 | - | 2 | 813 | ||
BC019247.1 | - | 2 | 822 | 4825175 | ||
U12978.2 | - | 2 | 918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
