siRNA Details
Gene Symbol: RNF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201365.1 | NP_001188294.1 | 5 | 832 | ||
NM_004290.4 | NP_004281.1 | 5 | 1046 | |||
NM_183398.2 | NP_899645.1 | 4 | 894 | |||
NM_183399.2 | NP_899646.1 | 4 | 658 | |||
NM_183400.2 | NP_899647.1 | 4 | 872 | |||
NM_183401.2 | NP_899648.1 | 5 | 852 | |||
XM_005268536.2 | XP_005268593.1 | 5 | 1149 | |||
XM_005268537.2 | XP_005268594.1 | 5 | 1193 | |||
XM_005268539.2 | XP_005268596.1 | 4 | 662 | |||
XM_005268540.3 | XP_005268597.1 | 4 | 818 | |||
XM_005268541.3 | XP_005268598.1 | 3 | 574 | |||
XM_011537714.2 | XP_011536016.1 | 4 | 663 | |||
XM_017010070.1 | XP_016865559.1 | 5 | 797 | |||
XM_017010071.1 | XP_016865560.1 | 4 | 1024 | |||
XM_017010072.1 | XP_016865561.1 | 4 | 984 | |||
GenBank mRNA | AB022663.1 | - | 5 | 820 | ||
AF060544.1 | - | 4 | 623 | |||
AK023698.1 | - | 4 | 622 | |||
AK023884.1 | - | 5 | 788 | |||
AK057868.1 | - | 4 | 670 | |||
AK129515.1 | - | 4 | 823 | |||
BC126185.1 | - | 5 | 791 | 8991901 | ||
BC144061.1 | - | 5 | 791 | 9052581 | ||
BX647955.1 | - | 5 | 819 | |||
CR456702.1 | - | 3 | 584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
