siRNA Details
Gene Symbol: H3F3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013699.2 | NP_001013721.2 | 1 | 322 | ||
GenBank mRNA | BC066906.1 | - | 1 | 322 | 5267388 | |
HQ873957.1 | - | 1 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
