siRNA Details
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 8 | 2348 | ||
XM_005266960.4 | XP_005267017.1 | 8 | 3578 | |||
XM_005266961.3 | XP_005267018.1 | 9 | 2398 | |||
XM_005266962.3 | XP_005267019.1 | 8 | 4618 | |||
XM_011535799.2 | XP_011534101.1 | 8 | 3660 | |||
GenBank mRNA | AK023349.1 | - | 3 | 1534 | ||
AK074311.1 | - | 2435 | ||||
BC040121.1 | - | 6 | 1993 | |||
BC046637.1 | - | 1 | 2096 | |||
BC047539.1 | - | 1 | 3257 | |||
BC065028.1 | - | 8 | 2272 | 6042215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
