siRNA Details
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 4 | 472 | ||
NR_104456.1 | 4 | 472 | ||||
XM_005266960.4 | XP_005267017.1 | 4 | 422 | |||
XM_005266961.3 | XP_005267018.1 | 4 | 422 | |||
XM_005266962.3 | XP_005267019.1 | 4 | 422 | |||
XM_011535798.2 | XP_011534100.1 | 4 | 422 | |||
XM_011535799.2 | XP_011534101.1 | 4 | 422 | |||
GenBank mRNA | AF514997.1 | - | 4 | 405 | ||
AK074311.1 | - | 427 | ||||
AK095994.1 | - | 4 | 428 | |||
AK301027.1 | - | 4 | 422 | |||
AK304244.1 | - | 4 | 404 | |||
BC040121.1 | - | 2 | 117 | |||
BC065028.1 | - | 4 | 398 | 6042215 | ||
BG567254.1 | - | 4 | 452 | |||
BX647097.1 | - | 4 | 422 | |||
CB989591.1 | - | 4 | 469 | |||
DC324759.1 | - | 4 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
