siRNA Details
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 3, 4 | 391 | ||
NR_104456.1 | 3, 4 | 391 | ||||
XM_005266960.4 | XP_005267017.1 | 3, 4 | 341 | |||
XM_005266961.3 | XP_005267018.1 | 3, 4 | 341 | |||
XM_005266962.3 | XP_005267019.1 | 3, 4 | 341 | |||
XM_011535798.2 | XP_011534100.1 | 3, 4 | 341 | |||
XM_011535799.2 | XP_011534101.1 | 3, 4 | 341 | |||
GenBank mRNA | AF514997.1 | - | 3, 4 | 324 | ||
AK074311.1 | - | 347 | ||||
AK301027.1 | - | 3, 4 | 341 | |||
AK304244.1 | - | 3, 4 | 323 | |||
BC040121.1 | - | 1, 2 | 36 | |||
BC065028.1 | - | 3, 4 | 317 | 6042215 | ||
BG567254.1 | - | 3, 4 | 371 | |||
BX647097.1 | - | 3, 4 | 341 | |||
CB989591.1 | - | 3, 4 | 388 | |||
DC324759.1 | - | 3, 4 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
