siRNA Details
Gene Symbol: CYB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190807.2 | NP_001177736.1 | 4 | 600 | ||
NM_001914.3 | NP_001905.1 | 6 | 654 | |||
NM_148923.3 | NP_683725.1 | 5 | 630 | |||
GenBank mRNA | AB209617.1 | - | 4 | 5709 | ||
AK026310.1 | - | 5 | 554 | |||
BC015182.1 | - | 5 | 558 | 3912638 | ||
BG196223.1 | - | 4 | 366 | |||
CA771478.1 | - | 4 | 566 | |||
M22865.1 | - | 5 | 541 | |||
M22976.1 | - | 3 | 968 | |||
M60174.1 | - | 6 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
