siRNA Details
Gene Symbol: CYB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190807.2 | NP_001177736.1 | 4 | 661 | ||
NM_001914.3 | NP_001905.1 | 6 | 715 | |||
NM_148923.3 | NP_683725.1 | 5 | 691 | |||
GenBank mRNA | AB209617.1 | - | 4 | 5770 | ||
AK026310.1 | - | 5 | 615 | |||
BC015182.1 | - | 5 | 619 | 3912638 | ||
BG196223.1 | - | 4 | 427 | |||
M22865.1 | - | 5 | 602 | |||
M22976.1 | - | 3 | 1029 | |||
M60174.1 | - | 6 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
