siRNA Details
Gene Symbol: CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204450.1 | NP_001191379.1 | 8 | 1204 | ||
NM_001204451.1 | NP_001191380.1 | 8 | 1204 | |||
NM_004748.4 | NP_004739.3 | 8 | 1444 | |||
NM_020739.3 | NP_065790.2 | 8 | 1204 | |||
GenBank mRNA | AB033080.2 | - | 8 | 989 | ||
AF212228.1 | - | 8 | 1120 | |||
AK022459.1 | - | 8 | 1000 | |||
AK292795.1 | - | 8 | 953 | |||
BC015203.1 | - | 8 | 1040 | |||
BC029398.1 | - | 1 | 71 | |||
BC034914.1 | - | 3 | 1362 | 5276323 | ||
BC039871.1 | - | 8 | 958 | 4385087 |
Gene Symbol: DYX1C1-CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037923.1 | 15 | 2322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
