siRNA Details
Gene Symbol: CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204450.1 | NP_001191379.1 | 7 | 1105 | ||
NM_001204451.1 | NP_001191380.1 | 7 | 1105 | |||
NM_004748.4 | NP_004739.3 | 7 | 1345 | |||
NM_020739.3 | NP_065790.2 | 7 | 1105 | |||
GenBank mRNA | AB033080.2 | - | 7 | 890 | ||
AF212228.1 | - | 7 | 1021 | |||
AK292795.1 | - | 7 | 854 | |||
BC015203.1 | - | 7 | 941 | |||
BC034914.1 | - | 2 | 1263 | 5276323 | ||
BC039871.1 | - | 7 | 859 | 4385087 |
Gene Symbol: DYX1C1-CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037923.1 | 14 | 2223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
