siRNA Details
Gene Symbol: NETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201465.1 | NP_001188394.1 | 3 | 407 | ||
NM_138966.3 | NP_620416.1 | 3 | 407 | |||
NM_138999.1 | NP_620552.1 | 3 | 222 | |||
XM_005266774.4 | XP_005266831.1 | 3 | 345 | |||
XM_017026017.1 | XP_016881506.1 | 3 | 269 | |||
XM_017026018.1 | XP_016881507.1 | 3 | 269 | |||
XM_017026019.1 | XP_016881508.1 | 3 | 429 | |||
XM_017026020.1 | XP_016881509.1 | 3 | 433 | |||
XM_017026021.1 | XP_016881510.1 | 2 | 95 | |||
XM_017026022.1 | XP_016881511.1 | 3 | 187 | |||
XM_017026023.1 | XP_016881512.1 | 3 | 187 | |||
XM_017026024.1 | XP_016881513.1 | 3 | 402 | |||
GenBank mRNA | AF448838.1 | - | 3 | 413 | ||
AF448839.1 | - | 3 | 222 | |||
AL834354.1 | - | 1 | 26 | |||
BC050329.1 | - | 3 | 409 | 4821317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
