siRNA Details
Gene Symbol: HIST1H4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003539.3 | NP_003530.1 | 1 | 25 | ||
GenBank mRNA | BC128104.1 | - | 1 | 25 | 40112084 | |
BC128105.1 | - | 1 | 24 | |||
BC143045.1 | - | 1 | 25 | 40112087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
