siRNA Details
Gene Symbol: C9orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252195.1 | NP_001239124.1 | 5 | 572 | ||
NM_032596.3 | NP_115985.2 | 7 | 1106 | |||
NM_147168.1 | NP_671697.1 | 4 | 648 | |||
NM_147169.2 | NP_671698.1 | 4 | 494 | |||
XM_005251616.4 | XP_005251673.1 | 7 | 4093 | |||
XM_005251618.4 | XP_005251675.1 | 5 | 640 | |||
XM_011518055.2 | XP_011516357.1 | 5 | 914 | |||
GenBank mRNA | AF367474.1 | - | 4 | 433 | ||
AK058087.1 | - | 7 | 1044 | |||
AK094751.1 | - | 2 | 2270 | |||
AK225981.1 | - | 4 | 566 | |||
BC022084.1 | - | 7 | 1091 | 4825995 | ||
BC029484.1 | - | 7 | 1106 | 5271301 | ||
CB854584.1 | - | 4 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
