siRNA Details
Gene Symbol: C9orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252195.1 | NP_001239124.1 | 2 | 105 | ||
NM_032596.3 | NP_115985.2 | 4 | 644 | |||
NM_147168.1 | NP_671697.1 | 1 | 186 | |||
NM_147169.2 | NP_671698.1 | 1 | 186 | |||
XM_005251616.4 | XP_005251673.1 | 4 | 3626 | |||
XM_005251618.4 | XP_005251675.1 | 2 | 173 | |||
XM_011518055.2 | XP_011516357.1 | 2 | 447 | |||
GenBank mRNA | AA338648.1 | - | 1 | 186 | ||
AF367474.1 | - | 1 | 125 | |||
AK058087.1 | - | 4 | 582 | |||
AK094751.1 | - | 1 | 175 | |||
AK225981.1 | - | 1 | 104 | |||
AW961020.1 | - | 1 | 186 | |||
BC022084.1 | - | 4 | 629 | 4825995 | ||
BC029484.1 | - | 4 | 644 | 5271301 | ||
BI561111.1 | - | 2 | 70 | |||
CB854584.1 | - | 1 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
