siRNA Details
Gene Symbol: TCF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193502.1 | NP_001180431.1 | 3 | 628 | ||
XM_017012940.1 | XP_016868429.1 | 1 | 1059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
