siRNA Details
Gene Symbol: OR2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012365.1 | NP_036497.1 | 1 | 320 | ||
GenBank mRNA | BC136858.1 | - | 1 | 354 | 9020849 | |
BC136860.1 | - | 1 | 354 | 9020851 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
