siRNA Details
Gene Symbol: NFKBID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321831.1 | NP_001308760.1 | 9, 10 | 1040 | ||
NM_139239.2 | NP_640332.1 | 9, 10 | 1203 | |||
XM_005259340.3 | XP_005259397.1 | 9, 10 | 1419 | |||
XM_011527419.2 | XP_011525721.1 | 9, 10 | 1091 | |||
XM_011527420.2 | XP_011525722.1 | 9, 10 | 1102 | |||
XM_011527421.2 | XP_011525723.1 | 8, 9 | 935 | |||
XM_011527424.2 | XP_011525726.1 | 9, 10 | 1047 | |||
XM_017027389.1 | XP_016882878.1 | 9, 10 | 1422 | |||
XM_017027390.1 | XP_016882879.1 | 9, 10 | 1401 | |||
XM_017027391.1 | XP_016882880.1 | 9, 10 | 1354 | |||
XM_017027392.1 | XP_016882881.1 | 9, 10 | 999 | |||
XM_017027393.1 | XP_016882882.1 | 9, 10 | 998 | |||
XM_017027394.1 | XP_016882883.1 | 7, 8 | 1246 | |||
XM_017027395.1 | XP_016882884.1 | 7, 8 | 1228 | |||
XM_017027397.1 | XP_016882886.1 | 7, 8 | 1185 | |||
GenBank mRNA | AF385433.1 | - | 9, 10 | 1093 | ||
AF385434.1 | - | 9, 10 | 1191 | |||
BC006273.2 | - | 3, 4 | 340 | |||
BC119646.1 | - | 6, 7 | 715 | 40115786 | ||
BC119647.1 | - | 6, 7 | 715 | 40115788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
