siRNA Details
Gene Symbol: NFKBID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321831.1 | NP_001308760.1 | 10 | 1093 | ||
NM_139239.2 | NP_640332.1 | 10 | 1256 | |||
XM_005259340.3 | XP_005259397.1 | 10 | 1472 | |||
XM_011527419.2 | XP_011525721.1 | 10 | 1144 | |||
XM_011527420.2 | XP_011525722.1 | 10 | 1155 | |||
XM_011527421.2 | XP_011525723.1 | 9 | 988 | |||
XM_011527424.2 | XP_011525726.1 | 10 | 1100 | |||
XM_017027389.1 | XP_016882878.1 | 10 | 1475 | |||
XM_017027390.1 | XP_016882879.1 | 10 | 1454 | |||
XM_017027391.1 | XP_016882880.1 | 10 | 1407 | |||
XM_017027392.1 | XP_016882881.1 | 10 | 1052 | |||
XM_017027393.1 | XP_016882882.1 | 10 | 1051 | |||
XM_017027394.1 | XP_016882883.1 | 8 | 1299 | |||
XM_017027395.1 | XP_016882884.1 | 8 | 1281 | |||
XM_017027397.1 | XP_016882886.1 | 8 | 1238 | |||
GenBank mRNA | AF385433.1 | - | 10 | 1146 | ||
AF385434.1 | - | 10 | 1244 | |||
BC006273.2 | - | 4 | 393 | |||
BC119646.1 | - | 7 | 768 | 40115786 | ||
BC119647.1 | - | 7 | 768 | 40115788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
