siRNA Details
Gene Symbol: GJB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005752.1 | NP_001005752.1 | 2 | 406 | ||
NM_024009.2 | NP_076872.1 | 2 | 863 | |||
GenBank mRNA | AF052692.1 | - | 1 | 1381 | ||
AK026139.1 | - | 2 | 862 | |||
AK312890.1 | - | 2 | 562 | |||
AL547036.3 | - | 2 | 624 | |||
BC012918.1 | - | 2 | 395 | 4500186 | ||
BC110640.1 | - | 1 | 1438 | 4780218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
