siRNA Details
Gene Symbol: ICAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001544.4 | NP_001535.1 | 2 | 497 | ||
NM_022377.3 | NP_071772.1 | 2 | 497 | |||
GenBank mRNA | BC029364.1 | - | 2 | 497 | 4659161 | |
L27670.1 | - | 2 | 457 | |||
L27671.1 | - | 2 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
