siRNA Details
Gene Symbol: IVNS1ABP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006469.4 | NP_006460.2 | 15 | 4035 | ||
XM_005244843.3 | XP_005244900.1 | 16 | 4132 | |||
XM_011509079.2 | XP_011507381.1 | 15 | 4066 | |||
XM_011509080.2 | XP_011507382.1 | 14 | 3900 | |||
XM_017000084.1 | XP_016855573.1 | 16 | 4153 | |||
GenBank mRNA | AF205218.1 | - | 15 | 3967 | ||
AL137640.1 | - | 1 | 1363 | |||
BX648273.1 | - | 8 | 3610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
