siRNA Details
Gene Symbol: IVNS1ABP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006469.4 | NP_006460.2 | 15 | 2362 | ||
XM_005244843.3 | XP_005244900.1 | 16 | 2459 | |||
XM_011509079.2 | XP_011507381.1 | 15 | 2393 | |||
XM_011509080.2 | XP_011507382.1 | 14 | 2227 | |||
XM_017000084.1 | XP_016855573.1 | 16 | 2480 | |||
GenBank mRNA | AB020657.1 | - | 15 | 2369 | ||
AF161553.1 | - | 15 | 2275 | |||
AF205218.1 | - | 15 | 2294 | |||
AJ012449.1 | - | 15 | 2276 | |||
AK000931.1 | - | 8 | 1002 | |||
AK001273.1 | - | 9 | 1193 | |||
AK001625.1 | - | 13 | 1700 | |||
AK001824.1 | - | 8 | 1080 | |||
AK023020.1 | - | 15 | 2111 | |||
AK092912.1 | - | 6 | 1362 | |||
AK093912.1 | - | 8 | 1879 | |||
AK292431.1 | - | 15 | 2112 | |||
BC067739.1 | - | 15 | 2262 | 5299055 | ||
BX648273.1 | - | 8 | 1939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
