siRNA Details
Gene Symbol: IVNS1ABP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006469.4 | NP_006460.2 | 15 | 2924 | ||
XM_005244843.3 | XP_005244900.1 | 16 | 3021 | |||
XM_011509079.2 | XP_011507381.1 | 15 | 2955 | |||
XM_011509080.2 | XP_011507382.1 | 14 | 2789 | |||
XM_017000084.1 | XP_016855573.1 | 16 | 3042 | |||
GenBank mRNA | AB020657.1 | - | 15 | 2931 | ||
AF205218.1 | - | 15 | 2856 | |||
AK000931.1 | - | 8 | 1564 | |||
AK001625.1 | - | 13 | 2262 | |||
AK001824.1 | - | 8 | 1642 | |||
AK023123.1 | - | 16 | 3005 | |||
AL137640.1 | - | 1 | 252 | |||
BX648273.1 | - | 8 | 2501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
