siRNA Details
Gene Symbol: TRIM64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136486.1 | NP_001129958.1 | 1 | 379 | ||
XM_011542585.2 | XP_011540887.1 | 2 | 468 |
Gene Symbol: TRIM64B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164397.1 | NP_001157869.1 | 1 | 379 | ||
XM_011542954.2 | XP_011541256.1 | 2 | 468 | |||
GenBank mRNA | JF968449.1 | - | 1 | 364 |
Gene Symbol: TRIM64C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206631.1 | NP_001193560.1 | 1 | 379 |
Gene Symbol: TRIM64EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968451.1 | - | 1 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
