siRNA Details
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 10 | 1609 | ||
NM_016032.3 | NP_057116.2 | 11 | 1589 | |||
GenBank mRNA | AB209807.1 | - | 10 | 1528 | ||
AF151847.1 | - | 11 | 1477 | |||
AK001112.1 | - | 11 | 1539 | |||
AK001424.1 | - | 8 | 923 | |||
AK001524.1 | - | 10 | 1340 | |||
AK096967.1 | - | 11 | 1240 | |||
AL161962.1 | - | 11 | 1466 | |||
AY358558.1 | - | 11 | 1521 | |||
AY952881.8 | - | 11 | 1561 | |||
BC000035.2 | - | 10 | 1483 | 3506601 | ||
BC003128.1 | - | 11 | 1527 | 2957911 | ||
BC006200.2 | - | 11 | 1523 | 2964425 | ||
BC012826.2 | - | 11 | 1523 | 2964425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
