siRNA Details
Gene Symbol: IGFL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207393.1 | NP_997276.1 | 3 | 177 | ||
XM_011526953.2 | XP_011525255.1 | 3 | 2604 | |||
XM_017026796.1 | XP_016882285.1 | 2 | 2715 | |||
XM_017026797.1 | XP_016882286.1 | 2 | 2715 | |||
GenBank mRNA | AY358434.1 | - | 3 | 177 | ||
AY672113.1 | - | 3 | 178 | |||
BC130593.1 | - | 3 | 176 | 40146624 | ||
BC130597.1 | - | 3 | 176 | 40146628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
