siRNA Details
Gene Symbol: OR8G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005198.1 | NP_001005198.1 | 1 | 517 | ||
XM_011542662.2 | XP_011540964.1 | 4 | 1071 | |||
XM_011542663.2 | XP_011540965.1 | 4 | 841 | |||
XM_011542664.2 | XP_011540966.1 | 3 | 710 | |||
XM_011542665.2 | XP_011540967.1 | 3 | 991 | |||
XM_011542666.2 | XP_011540968.1 | 3 | 761 | |||
XM_011542667.2 | XP_011540969.1 | 2 | 630 | |||
GenBank mRNA | BC136820.1 | - | 1 | 540 | 9020811 | |
BC136822.1 | - | 1 | 540 | 9020813 |
Gene Symbol: OR8G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002905.1 | NP_001002905.1 | 1 | 412 | ||
NR_045681.1 | 1 | 412 | ||||
GenBank mRNA | X89672.1 | - | 1 | 38 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
