siRNA Details
Gene Symbol: OR8G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005198.1 | NP_001005198.1 | 1 | 947 | ||
XM_011542662.2 | XP_011540964.1 | 4 | 1501 | |||
XM_011542663.2 | XP_011540965.1 | 4 | 1271 | |||
XM_011542664.2 | XP_011540966.1 | 3 | 1140 | |||
XM_011542665.2 | XP_011540967.1 | 3 | 1421 | |||
XM_011542666.2 | XP_011540968.1 | 3 | 1191 | |||
XM_011542667.2 | XP_011540969.1 | 2 | 1060 | |||
GenBank mRNA | BC136820.1 | - | 1 | 970 | 9020811 |
Gene Symbol: OR8G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136639.1 | - | 1 | 935 | 9020629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
