siRNA Details
Gene Symbol: LRRC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023126.1 | XP_016878615.1 | 6 | 1395 | ||
XM_017023127.1 | XP_016878616.1 | 6 | 1646 | |||
XM_017023128.1 | XP_016878617.1 | 6 | 1451 | |||
XM_017023129.1 | XP_016878618.1 | 6 | 2537 | |||
XM_017023130.1 | XP_016878619.1 | 6 | 1119 | |||
XM_017023131.1 | XP_016878620.1 | 6 | 978 | |||
XM_017023132.1 | XP_016878621.1 | 6 | 1281 | |||
XM_017023134.1 | XP_016878623.1 | 6 | 1086 | |||
GenBank mRNA | AF176701.1 | - | 6 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
