siRNA Details
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006945.1 | NP_001006946.1 | 2 | 496 | ||
NM_018372.3 | NP_060842.3 | 3 | 2024 | |||
XM_005271029.3 | XP_005271086.1 | 3 | 2283 | |||
XM_011541768.1 | XP_011540070.1 | 3 | 2024 | |||
XM_017001768.1 | XP_016857257.1 | 3 | 2024 | |||
XM_017001769.1 | XP_016857258.1 | 3 | 2283 | |||
GenBank mRNA | AK001826.1 | - | 2 | 602 | ||
AK002131.1 | - | 2 | 965 | |||
AL834466.1 | - | 2 | 942 | |||
AY190122.1 | - | 3 | 2014 | |||
BC008115.1 | - | 2 | 1166 | 2900378 | ||
BU664417.1 | - | 2 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
