siRNA Details
Gene Symbol: SYN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003178.5 | NP_003169.2 | 2 | 612 | ||
NM_133625.4 | NP_598328.1 | 2 | 612 | |||
XM_006713311.3 | XP_006713374.1 | 2 | 606 | |||
GenBank mRNA | AB209088.1 | - | 3 | 530 | ||
AF077671.1 | - | 2 | 441 | |||
AF077737.1 | - | 2 | 441 | |||
AK094464.1 | - | 3 | 380 | |||
BC051307.1 | - | 2 | 570 | 5802100 | ||
U40215.1 | - | 2 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
