siRNA Details
Gene Symbol: SYN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003178.5 | NP_003169.2 | 7 | 1151 | ||
NM_133625.4 | NP_598328.1 | 7 | 1151 | |||
XM_006713311.3 | XP_006713374.1 | 7 | 1145 | |||
XM_006713312.3 | XP_006713375.1 | 4 | 1353 | |||
XM_006713313.2 | XP_006713376.1 | 4 | 491 | |||
XM_017007087.1 | XP_016862576.1 | 3 | 563 | |||
GenBank mRNA | AB209088.1 | - | 8 | 1069 | ||
AF077671.1 | - | 7 | 980 | |||
AF077737.1 | - | 7 | 980 | |||
AK091286.1 | - | 4 | 578 | |||
AK094464.1 | - | 8 | 919 | |||
AK295431.1 | - | 4 | 367 | |||
AK307469.1 | - | 3 | 232 | |||
BC051307.1 | - | 7 | 1109 | 5802100 | ||
U40215.1 | - | 7 | 1015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
