siRNA Details
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 14 | 1818 | ||
NM_199179.3 | NP_954648.3 | 13 | 1668 | |||
NM_199180.3 | NP_954649.3 | 14 | 1960 | |||
XM_011527362.1 | XP_011525664.1 | 15 | 2020 | |||
XM_011527363.1 | XP_011525665.1 | 15 | 2001 | |||
XM_011527365.1 | XP_011525667.1 | 14 | 1870 | |||
XM_011527366.1 | XP_011525668.1 | 13 | 2131 | |||
XM_017027348.1 | XP_016882837.1 | 13 | 2422 | |||
GenBank mRNA | AB593116.1 | - | 14 | 1924 | ||
AK307865.1 | - | 13 | 2176 | |||
AL136654.1 | - | 14 | 1944 | |||
AY305301.1 | - | 14 | 1762 | |||
AY305302.1 | - | 13 | 1612 | |||
AY358742.1 | - | 14 | 1946 | |||
BC007312.1 | - | 5 | 493 | |||
BC064925.1 | - | 17 | 1883 | 6141879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
