siRNA Details
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 13 | 1742 | ||
NM_199179.3 | NP_954648.3 | 12 | 1592 | |||
NM_199180.3 | NP_954649.3 | 13 | 1884 | |||
XM_011527362.1 | XP_011525664.1 | 14 | 1944 | |||
XM_011527363.1 | XP_011525665.1 | 14 | 1925 | |||
XM_011527365.1 | XP_011525667.1 | 13 | 1794 | |||
XM_011527366.1 | XP_011525668.1 | 12 | 2055 | |||
XM_017027348.1 | XP_016882837.1 | 12 | 2346 | |||
GenBank mRNA | AB593116.1 | - | 13 | 1848 | ||
AK307865.1 | - | 12 | 2100 | |||
AL136654.1 | - | 13 | 1868 | |||
AY305301.1 | - | 13 | 1686 | |||
AY305302.1 | - | 12 | 1536 | |||
AY358742.1 | - | 13 | 1870 | |||
BC007312.1 | - | 4 | 417 | |||
BC064925.1 | - | 16 | 1807 | 6141879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
