siRNA Details
Gene Symbol: PMCHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003922.1 | 2 | 309 | |||
GenBank mRNA | AY008412.1 | - | 2 | 309 | ||
AY008413.1 | - | 2 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
