siRNA Details
Gene Symbol: TAF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243156.1 | NP_001230085.1 | 15 | 3782 | ||
NM_001243157.1 | NP_001230086.1 | 12 | 3416 | |||
NM_001243158.1 | NP_001230087.1 | 11 | 3277 | |||
NM_001243159.1 | NP_001230088.1 | 14 | 3749 | |||
NM_001243160.1 | NP_001230089.1 | 12 | 3594 | |||
NM_005679.3 | NP_005670.3 | 14 | 3860 | |||
NM_139353.2 | NP_647610.2 | 15 | 3618 | |||
XM_005256226.3 | XP_005256283.1 | 14 | 3783 | |||
XM_005256227.3 | XP_005256284.1 | 14 | 3694 | |||
XM_006721325.3 | XP_006721388.1 | 15 | 3875 | |||
XM_006721326.3 | XP_006721389.1 | 16 | 3797 | |||
XM_017023845.1 | XP_016879334.1 | 15 | 3705 | |||
XM_017023846.1 | XP_016879335.1 | 13 | 3644 | |||
XM_017023847.1 | XP_016879336.1 | 15 | 3616 | |||
XM_017023848.1 | XP_016879337.1 | 9 | 3467 | |||
GenBank mRNA | AB209594.1 | - | 13 | 4558 | ||
AK304261.1 | - | 14 | 3731 | |||
BC028131.1 | - | 15 | 3597 | 5205601 | ||
L39059.1 | - | 14 | 3801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
