siRNA Details
Gene Symbol: TAF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243156.1 | NP_001230085.1 | 9 | 1140 | ||
NM_001243157.1 | NP_001230086.1 | 6 | 774 | |||
NM_001243158.1 | NP_001230087.1 | 5 | 635 | |||
NM_001243159.1 | NP_001230088.1 | 9 | 979 | |||
NM_001243160.1 | NP_001230089.1 | 7 | 843 | |||
NM_005679.3 | NP_005670.3 | 8 | 1218 | |||
NM_139353.2 | NP_647610.2 | 9 | 979 | |||
XM_005256226.3 | XP_005256283.1 | 8 | 1141 | |||
XM_005256227.3 | XP_005256284.1 | 8 | 1052 | |||
XM_006721325.3 | XP_006721388.1 | 9 | 1233 | |||
XM_006721326.3 | XP_006721389.1 | 10 | 1155 | |||
XM_017023845.1 | XP_016879334.1 | 9 | 1063 | |||
XM_017023846.1 | XP_016879335.1 | 7 | 1002 | |||
XM_017023847.1 | XP_016879336.1 | 9 | 974 | |||
XM_017023848.1 | XP_016879337.1 | 3 | 825 | |||
GenBank mRNA | AB209594.1 | - | 9 | 1090 | ||
AK097031.1 | - | 7 | 812 | |||
AK299060.1 | - | 6 | 737 | |||
AK299347.1 | - | 9 | 1111 | |||
AK304261.1 | - | 9 | 961 | |||
AK304739.1 | - | 10 | 1092 | |||
AK315982.1 | - | 5 | 622 | |||
BC028131.1 | - | 9 | 958 | 5205601 | ||
L39059.1 | - | 8 | 1160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
