siRNA Details
Gene Symbol: MPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015134.3 | NP_055949.2 | 23 | 3832 | ||
NM_201274.3 | NP_958431.2 | 24 | 3895 | |||
XM_005256563.4 | XP_005256620.1 | 24 | 7944 | |||
XM_005256564.4 | XP_005256621.3 | 22 | 4023 | |||
XM_011523761.2 | XP_011522063.1 | 24 | 8315 | |||
XM_011523762.2 | XP_011522064.1 | 25 | 8361 | |||
XM_011523763.2 | XP_011522065.1 | 23 | 8184 | |||
XM_011523764.2 | XP_011522066.1 | 23 | 8178 | |||
XM_011523765.2 | XP_011522067.1 | 24 | 8241 | |||
XM_011523766.2 | XP_011522068.2 | 23 | 4491 | |||
XM_011523767.2 | XP_011522069.2 | 24 | 4554 | |||
XM_017024393.1 | XP_016879882.1 | 23 | 4434 | |||
GenBank mRNA | AB020671.2 | - | 9 | 4836 | ||
AB098507.1 | - | 23 | 3796 | |||
AK025604.1 | - | 10 | 4002 | |||
AK092287.1 | - | 19 | 3273 | |||
AL834513.1 | - | 23 | 3692 | |||
BC009982.2 | - | 18 | 3155 | |||
BC012426.1 | - | 5 | 1081 | |||
BC021058.1 | - | 11 | 1803 | |||
BC021070.1 | - | 11 | 1803 | |||
BC075847.1 | - | 19 | 3229 | |||
D23673.1 | - | 9 | 1709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
