siRNA Details
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201362.1 | NP_001188291.1 | 5 | 1410 | ||
NM_001201363.1 | NP_001188292.1 | 7 | 954 | |||
NM_001318884.1 | NP_001305813.1 | 7 | 961 | |||
NM_001318885.1 | NP_001305814.1 | 5 | 810 | |||
NM_001318886.1 | NP_001305815.1 | 7 | 948 | |||
NM_001318887.1 | NP_001305816.1 | 6 | 1290 | |||
NM_001318888.1 | NP_001305817.1 | 5 | 747 | |||
NM_004242.3 | NP_004233.1 | 6 | 861 | |||
NM_138730.2 | NP_620058.1 | 6 | 820 | |||
GenBank mRNA | AF274949.1 | - | 6 | 780 | ||
AF401520.1 | - | 6 | 711 | |||
AK124181.1 | - | 1 | 1749 | |||
AY043282.1 | - | 6 | 828 | |||
BC009529.2 | - | 6 | 748 | 3890883 | ||
BM927225.1 | - | 7 | 824 | |||
BX648085.1 | - | 1 | 4651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
