siRNA Details
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201362.1 | NP_001188291.1 | 5 | 1273 | ||
NM_001201363.1 | NP_001188292.1 | 7 | 817 | |||
NM_001318884.1 | NP_001305813.1 | 7 | 824 | |||
NM_001318885.1 | NP_001305814.1 | 5 | 673 | |||
NM_001318886.1 | NP_001305815.1 | 7 | 811 | |||
NM_001318887.1 | NP_001305816.1 | 6 | 1153 | |||
NM_001318888.1 | NP_001305817.1 | 5 | 610 | |||
NM_004242.3 | NP_004233.1 | 6 | 724 | |||
NM_138730.2 | NP_620058.1 | 6 | 683 | |||
GenBank mRNA | AF274949.1 | - | 6 | 643 | ||
AF401520.1 | - | 6 | 574 | |||
AK124181.1 | - | 1 | 1612 | |||
AY043282.1 | - | 6 | 691 | |||
BC009529.2 | - | 6 | 611 | 3890883 | ||
BM927225.1 | - | 7 | 687 | |||
BX648085.1 | - | 1 | 4514 | |||
CF553005.1 | - | 7 | 688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
