siRNA Details
Gene Symbol: LOC107986803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012930.1 | XP_016868419.1 | 1 | 1260 |
Gene Symbol: LOC100132217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128836.1 | - | 1 | 1309 |
Gene Symbol: LOC100996365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250737.3 | XP_005250794.1 | 1 | 644 |
Gene Symbol: LOC107986804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012931.1 | XP_016868420.1 | 1 | 1225 |
Gene Symbol: LOC401357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130727.1 | 1 | 603 | |||
GenBank mRNA | AK125790.1 | - | 1 | 602 | ||
AK128114.1 | - | 1 | 344 | |||
BC137417.1 | - | 1 | 338 | |||
JX508854.1 | - | 1 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
