siRNA Details
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008395.3 | NP_001008396.1 | 4 | 473 | ||
NM_001318236.1 | NP_001305165.1 | 4 | 490 | |||
NM_001318237.1 | NP_001305166.1 | 3 | 392 | |||
NR_134526.1 | 4 | 459 | ||||
NR_134527.1 | 5 | 515 | ||||
NR_134528.1 | 4 | 472 | ||||
NR_134529.1 | 4 | 452 | ||||
NR_134530.1 | 4 | 489 | ||||
XM_017012198.1 | XP_016867687.1 | 6 | 860 | |||
XM_017012199.1 | XP_016867688.1 | 6 | 877 | |||
GenBank mRNA | BC063401.1 | - | 4 | 453 | 5194116 | |
BC105609.1 | - | 4 | 463 | 30321610 | ||
BC130553.1 | - | 4 | 420 | 40146584 | ||
BC130559.1 | - | 4 | 420 | 40146590 | ||
BE733059.1 | - | 3 | 382 | |||
BG706819.1 | - | 4 | 473 | |||
BG715626.1 | - | 4 | 470 | |||
BG819624.1 | - | 4 | 433 | |||
BI598677.1 | - | 4 | 476 | |||
BI771662.1 | - | 3 | 378 | |||
DB498044.1 | - | 4 | 468 | |||
DB539334.2 | - | 3 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
