siRNA Details
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008395.3 | NP_001008396.1 | 3 | 258 | ||
NM_001318236.1 | NP_001305165.1 | 3 | 258 | |||
NM_001318237.1 | NP_001305166.1 | 2 | 177 | |||
NR_134526.1 | 3 | 244 | ||||
NR_134527.1 | 4 | 300 | ||||
NR_134528.1 | 3 | 257 | ||||
NR_134529.1 | 3 | 237 | ||||
NR_134530.1 | 3 | 257 | ||||
XM_017012198.1 | XP_016867687.1 | 5 | 645 | |||
XM_017012199.1 | XP_016867688.1 | 5 | 645 | |||
GenBank mRNA | AJ714519.1 | - | 3 | 238 | ||
BC063401.1 | - | 3 | 238 | 5194116 | ||
BC105609.1 | - | 3 | 248 | 30321610 | ||
BC130553.1 | - | 3 | 205 | 40146584 | ||
BC130559.1 | - | 3 | 205 | 40146590 | ||
BE733059.1 | - | 2 | 168 | |||
BF978089.1 | - | 257 | ||||
BG706819.1 | - | 3 | 241 | |||
BG715626.1 | - | 3 | 254 | |||
BG819624.1 | - | 3 | 218 | |||
BI598677.1 | - | 3 | 261 | |||
BI771662.1 | - | 2 | 163 | |||
CU450474.1 | - | 3 | 311 | |||
DB498044.1 | - | 3 | 236 | |||
DB539334.2 | - | 2 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
