siRNA Details
Gene Symbol: FMNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175736.4 | NP_783863.4 | 17 | 2143 | ||
NM_198900.2 | NP_944489.2 | 16 | 1990 | |||
XM_005269218.2 | XP_005269275.1 | 17 | 2122 | |||
XM_011538968.2 | XP_011537270.1 | 17 | 2612 | |||
XM_011538969.2 | XP_011537271.1 | 17 | 2611 | |||
XM_011538970.2 | XP_011537272.1 | 17 | 2611 | |||
XM_011538971.2 | XP_011537273.1 | 17 | 2187 | |||
XM_011538972.2 | XP_011537274.1 | 16 | 2457 | |||
XM_011538973.2 | XP_011537275.2 | 13 | 1602 | |||
XM_011538974.2 | XP_011537276.1 | 18 | 1944 | |||
GenBank mRNA | AB095934.2 | - | 17 | 2143 | ||
AK127200.1 | - | 4 | 616 | |||
AK128195.1 | - | 16 | 1970 | |||
AL162062.1 | - | 1 | 100 | |||
BC159100.1 | - | 17 | 2143 | 8860524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
