siRNA Details
Gene Symbol: FMNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175736.4 | NP_783863.4 | 19 | 2515 | ||
NM_198900.2 | NP_944489.2 | 18 | 2362 | |||
XM_005269218.2 | XP_005269275.1 | 19 | 2494 | |||
XM_011538968.2 | XP_011537270.1 | 19 | 2984 | |||
XM_011538969.2 | XP_011537271.1 | 19 | 2983 | |||
XM_011538970.2 | XP_011537272.1 | 19 | 2983 | |||
XM_011538971.2 | XP_011537273.1 | 19 | 2559 | |||
XM_011538972.2 | XP_011537274.1 | 18 | 2829 | |||
XM_011538973.2 | XP_011537275.2 | 15 | 1974 | |||
XM_011538974.2 | XP_011537276.1 | 20 | 2316 | |||
GenBank mRNA | AB095934.2 | - | 19 | 2515 | ||
AK127200.1 | - | 6 | 988 | |||
AK128195.1 | - | 18 | 2342 | |||
AL162062.1 | - | 3 | 472 | |||
BC033181.1 | - | 3 | 299 | |||
BC159100.1 | - | 19 | 2515 | 8860524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
